siRNA Details
| Gene Symbol: OR56B4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005181.2 | NP_001005181.1 | 1 | 573 | ||
| GenBank mRNA | BC136879.1 | - | 1 | 573 | 9020870 | |
| BC136880.1 | - | 1 | 573 | 9020871 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR56B4| Gene Symbol: OR56B4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005181.2 | NP_001005181.1 | 1 | 573 | ||
| GenBank mRNA | BC136879.1 | - | 1 | 573 | 9020870 | |
| BC136880.1 | - | 1 | 573 | 9020871 | ||