siRNA Details
| Gene Symbol: OR6C1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005182.1 | NP_001005182.1 | 1 | 74 | ||
| GenBank mRNA | BC136957.1 | - | 1 | 113 | 9020949 | |
| BC136965.1 | - | 1 | 112 | 9020957 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR6C1| Gene Symbol: OR6C1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005182.1 | NP_001005182.1 | 1 | 74 | ||
| GenBank mRNA | BC136957.1 | - | 1 | 113 | 9020949 | |
| BC136965.1 | - | 1 | 112 | 9020957 | ||