siRNA Details
| Gene Symbol: Olfr1024 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005230.2 | NP_001005230.2 | 1 | 810 | ||
| GenBank mRNA | BC119447.1 | - | 1 | 810 | 40044257 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1024| Gene Symbol: Olfr1024 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005230.2 | NP_001005230.2 | 1 | 810 | ||
| GenBank mRNA | BC119447.1 | - | 1 | 810 | 40044257 | |