siRNA Details
| Gene Symbol: OR4C12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005270.4 | NP_001005270.3 | 1 | 421 | ||
| GenBank mRNA | BC136850.1 | - | 1 | 421 | 9020841 | |
| BC136851.1 | - | 1 | 421 | 9020842 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR4C12| Gene Symbol: OR4C12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005270.4 | NP_001005270.3 | 1 | 421 | ||
| GenBank mRNA | BC136850.1 | - | 1 | 421 | 9020841 | |
| BC136851.1 | - | 1 | 421 | 9020842 | ||