siRNA Details
| Gene Symbol: Klk1c12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005382.1 | NP_001005382.1 | 4 | 561 | ||
| Gene Symbol: LOC687880 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006229168.1 | XP_006229230.1 | 567 | |||

Klk1c12
Genomic Map