siRNA Details
| Gene Symbol: Stmnd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005422.1 | NP_001005422.1 | 5 | 930 | ||
| GenBank mRNA | BC064117.1 | - | 5 | 930 | 3418830 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Stmnd1| Gene Symbol: Stmnd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005422.1 | NP_001005422.1 | 5 | 930 | ||
| GenBank mRNA | BC064117.1 | - | 5 | 930 | 3418830 | |