siRNA Details
| Gene Symbol: Mreg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005423.2 | NP_001005423.1 | 5 | 1858 | ||
| GenBank mRNA | AY628210.1 | - | 5 | 1662 | ||
| BC068125.1 | - | 5 | 1857 | 6828150 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mreg| Gene Symbol: Mreg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005423.2 | NP_001005423.1 | 5 | 1858 | ||
| GenBank mRNA | AY628210.1 | - | 5 | 1662 | ||
| BC068125.1 | - | 5 | 1857 | 6828150 | ||