siRNA Details
| Gene Symbol: ZBTB40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001083621.1 | NP_001077090.1 | 13, 14 | 3187 | ||
| NM_014870.3 | NP_055685.3 | 12, 13 | 2899 | |||
| XM_011542499.2 | XP_011540801.1 | 12, 13 | 2922 | |||
| XM_011542500.2 | XP_011540802.1 | 5, 6 | 2172 | |||
| XM_017003003.1 | XP_016858492.1 | 10, 11 | 2563 | |||
| GenBank mRNA | AB007947.2 | - | 12, 13 | 2893 | ||
| AK314747.1 | - | 2, 3 | 601 | |||
| BC114607.1 | - | 10, 11 | 2435 | 40027579 | ||

ZBTB40
Genomic Map