siRNA Details
Gene Symbol: Zfp663 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005425.1 | NP_001005425.1 | 5 | 771 | ||
GenBank mRNA | AK141109.1 | - | 6 | 911 | ||
BC067000.1 | - | 5 | 771 | 5705933 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zfp663 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005425.1 | NP_001005425.1 | 5 | 771 | ||
GenBank mRNA | AK141109.1 | - | 6 | 911 | ||
BC067000.1 | - | 5 | 771 | 5705933 |