siRNA Details
Gene Symbol: PLCXD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005473.2 | NP_001005473.1 | 3 | 4895 | ||
GenBank mRNA | AK124638.1 | - | 1 | 1120 | ||
BX648329.1 | - | 4 | 4706 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: PLCXD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005473.2 | NP_001005473.1 | 3 | 4895 | ||
GenBank mRNA | AK124638.1 | - | 1 | 1120 | ||
BX648329.1 | - | 4 | 4706 |