siRNA Details
Gene Symbol: OR4K5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005483.1 | NP_001005483.1 | 1 | 893 | ||
GenBank mRNA | BC140947.1 | - | 1 | 918 | 9056759 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: OR4K5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005483.1 | NP_001005483.1 | 1 | 893 | ||
GenBank mRNA | BC140947.1 | - | 1 | 918 | 9056759 |