siRNA Details
| Gene Symbol: OR10AG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005491.1 | NP_001005491.1 | 1 | 744 | ||
| GenBank mRNA | BC136890.1 | - | 1 | 795 | 9020881 | |
| BC136891.1 | - | 1 | 795 | 9020882 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR10AG1| Gene Symbol: OR10AG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005491.1 | NP_001005491.1 | 1 | 744 | ||
| GenBank mRNA | BC136890.1 | - | 1 | 795 | 9020881 | |
| BC136891.1 | - | 1 | 795 | 9020882 | ||