siRNA Details
| Gene Symbol: Lmtk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005511.3 | NP_001005511.2 | 10 | 1062 | ||
| NM_001290990.1 | NP_001277919.1 | 9 | 1217 | |||
| GenBank mRNA | AB288873.1 | - | 10 | 1056 | ||
| BC057032.1 | - | 9 | 950 | |||
| BC059845.1 | - | 10 | 1267 | |||
| BC083185.1 | - | 9 | 1217 | 6855048 | ||
| BC094377.1 | - | 10 | 1062 | 30536822 | ||

Lmtk3
Genomic Map