siRNA Details
| Gene Symbol: N4bp2l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005533.1 | NP_001005533.1 | 3, 4 | 1458 | ||
| XM_006248735.2 | XP_006248797.1 | 3, 4 | 1444 | |||
| XM_006248736.2 | XP_006248798.1 | 3, 4 | 1469 | |||
| GenBank mRNA | BC081998.1 | - | 3, 4 | 1458 | 7131024 | |

N4bp2l2
Genomic Map