siRNA Details
| Gene Symbol: Ik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005537.1 | NP_001005537.1 | 9 | 811 | ||
| GenBank mRNA | BC081870.1 | - | 9 | 811 | 7106138 | |
| FQ212525.1 | - | 9 | 765 | |||
| FQ214775.1 | - | 6 | 521 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ik| Gene Symbol: Ik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005537.1 | NP_001005537.1 | 9 | 811 | ||
| GenBank mRNA | BC081870.1 | - | 9 | 811 | 7106138 | |
| FQ212525.1 | - | 9 | 765 | |||
| FQ214775.1 | - | 6 | 521 | |||