siRNA Details
| Gene Symbol: Fam160a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005538.2 | NP_001005538.2 | 12 | 3346 | ||
| XM_006229931.2 | XP_006229993.1 | 12 | 3338 | |||
| XM_006229932.2 | XP_006229994.1 | 12 | 2785 | |||
| GenBank mRNA | BC082011.1 | - | 12 | 2738 | 7132039 | |

Fam160a2
Genomic Map