siRNA Details
| Gene Symbol: Tspan3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005547.1 | NP_001005547.1 | 2 | 222 | ||
| GenBank mRNA | BC082102.1 | - | 2 | 222 | 7134588 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tspan3| Gene Symbol: Tspan3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005547.1 | NP_001005547.1 | 2 | 222 | ||
| GenBank mRNA | BC082102.1 | - | 2 | 222 | 7134588 | |