siRNA Details
| Gene Symbol: Tinag | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005549.1 | NP_001005549.1 | 1 | 181 | ||
| GenBank mRNA | BC081887.1 | - | 1 | 181 | 7109073 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tinag| Gene Symbol: Tinag | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005549.1 | NP_001005549.1 | 1 | 181 | ||
| GenBank mRNA | BC081887.1 | - | 1 | 181 | 7109073 | |