siRNA Details
| Gene Symbol: MGC95208 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005552.1 | NP_001005552.1 | 3 | 932 | ||
| GenBank mRNA | BC082055.1 | - | 3 | 932 | 7132244 | |
| BC091244.1 | - | 3 | 927 | 7365330 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MGC95208| Gene Symbol: MGC95208 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005552.1 | NP_001005552.1 | 3 | 932 | ||
| GenBank mRNA | BC082055.1 | - | 3 | 932 | 7132244 | |
| BC091244.1 | - | 3 | 927 | 7365330 | ||