siRNA Details
| Gene Symbol: Git2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005553.1 | NP_001005553.1 | 14 | 1436 | ||
| XM_006249442.2 | XP_006249504.1 | 14 | 1481 | |||
| XM_006249445.2 | XP_006249507.1 | 14 | 1484 | |||
| GenBank mRNA | BC081967.1 | - | 14 | 1436 | 7125406 | |

Git2
Genomic Map