siRNA Details
| Gene Symbol: Mavs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005556.1 | NP_001005556.1 | 7 | 1436 | ||
| XM_006235034.2 | XP_006235096.1 | 7 | 1304 | |||
| XM_006235035.2 | XP_006235097.1 | 7 | 2018 | |||
| XM_006235036.2 | XP_006235098.1 | 8 | 1337 | |||
| XM_006235037.2 | XP_006235099.1 | 6 | 1340 | |||
| XM_008762179.1 | XP_008760401.1 | 7 | 2511 | |||
| XM_008762180.1 | XP_008760402.1 | 7 | 1324 | |||
| GenBank mRNA | BC081869.1 | - | 7 | 1436 | 7106026 | |

Mavs
Genomic Map