siRNA Details
| Gene Symbol: PDGFRL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006207.2 | NP_006198.1 | 5 | 964 | ||
| GenBank mRNA | AK289450.1 | - | 5 | 964 | ||
| BC010927.1 | - | 4 | 535 | 4278882 | ||
| CR541952.1 | - | 4 | 519 | |||
| D37965.1 | - | 4 | 580 | |||

PDGFRL
Genomic Map