siRNA Details
| Gene Symbol: Fam151a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005558.1 | NP_001005558.1 | 2, 3 | 315 | ||
| GenBank mRNA | BC081977.1 | - | 2, 3 | 315 | 7127557 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam151a| Gene Symbol: Fam151a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005558.1 | NP_001005558.1 | 2, 3 | 315 | ||
| GenBank mRNA | BC081977.1 | - | 2, 3 | 315 | 7127557 | |