siRNA Details
| Gene Symbol: Parl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005767.4 | NP_001005767.1 | 2 | 254 | ||
| GenBank mRNA | AK039142.1 | - | 2 | 233 | ||
| AK150980.1 | - | 2 | 262 | |||
| AK151652.1 | - | 2 | 253 | |||
| AK179606.1 | - | 2 | 119 | |||
| AK181973.1 | - | 2 | 120 | |||
| AK207637.1 | - | 2 | 263 | |||
| AK216516.1 | - | 2 | 119 | |||
| BC083153.1 | - | 2 | 254 | 30728006 | ||

Parl
Genomic Map