siRNA Details
| Gene Symbol: Zfp69 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005788.3 | NP_001005788.2 | 6 | 1135 | ||
| GenBank mRNA | BC066020.1 | - | 5 | 734 | 5717218 | |
| U29497.1 | - | 3 | 283 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp69| Gene Symbol: Zfp69 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005788.3 | NP_001005788.2 | 6 | 1135 | ||
| GenBank mRNA | BC066020.1 | - | 5 | 734 | 5717218 | |
| U29497.1 | - | 3 | 283 | |||