siRNA Details
| Gene Symbol: Aga | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005847.2 | NP_001005847.1 | 2 | 312 | ||
| NM_001205054.1 | NP_001191983.1 | 2 | 312 | |||
| GenBank mRNA | AA396103.1 | - | 2 | 157 | ||
| BC048230.1 | - | 2 | 1193 | |||
| BC132227.1 | - | 2 | 287 | 40130504 | ||
| BC132229.1 | - | 2 | 287 | 40130506 | ||
| BC145297.1 | - | 2 | 287 | 9053851 | ||
| BQ571087.1 | - | 2 | 204 | |||
| CD541877.1 | - | 2 | 288 | |||
| S81393.1 | - | 2 | 229 | |||

Aga
Genomic Map