siRNA Details
| Gene Symbol: Psmd12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005875.1 | NP_001005875.1 | 11 | 1234 | ||
| GenBank mRNA | BC083758.1 | - | 11 | 1234 | 7192849 | |
| FQ218292.1 | - | 5 | 461 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Psmd12| Gene Symbol: Psmd12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005875.1 | NP_001005875.1 | 11 | 1234 | ||
| GenBank mRNA | BC083758.1 | - | 11 | 1234 | 7192849 | |
| FQ218292.1 | - | 5 | 461 | |||