siRNA Details
Gene Symbol: Hn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005876.1 | NP_001005876.1 | 5 | 1198 | ||
GenBank mRNA | BC079311.1 | - | 5 | 1198 | 7134794 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Hn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005876.1 | NP_001005876.1 | 5 | 1198 | ||
GenBank mRNA | BC079311.1 | - | 5 | 1198 | 7134794 |