siRNA Details
| Gene Symbol: Ffar2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005877.1 | NP_001005877.1 | 1 | 592 | ||
| XM_006228821.2 | XP_006228883.1 | 2 | 1344 | |||
| XM_006228822.2 | XP_006228884.1 | 3 | 938 | |||
| XM_008759133.1 | XP_008757355.1 | 3 | 1038 | |||
| XM_008759134.1 | XP_008757356.1 | 3 | 999 | |||
| XM_008759135.1 | XP_008757357.1 | 3 | 716 | |||
| GenBank mRNA | AB106675.1 | - | 1 | 592 | ||

Ffar2
Genomic Map