siRNA Details
| Gene Symbol: Gfpt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005879.1 | NP_001005879.1 | 13 | 1312 | ||
| XM_006236823.2 | XP_006236885.1 | 14 | 1438 | |||
| GenBank mRNA | BC083889.1 | - | 13 | 1312 | 7132442 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gfpt1| Gene Symbol: Gfpt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005879.1 | NP_001005879.1 | 13 | 1312 | ||
| XM_006236823.2 | XP_006236885.1 | 14 | 1438 | |||
| GenBank mRNA | BC083889.1 | - | 13 | 1312 | 7132442 | |