siRNA Details
Gene Symbol: Pi4k2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005883.1 | NP_001005883.1 | 6, 7 | 1033 | ||
GenBank mRNA | BC083668.1 | - | 6, 7 | 1033 | 7135573 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Pi4k2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005883.1 | NP_001005883.1 | 6, 7 | 1033 | ||
GenBank mRNA | BC083668.1 | - | 6, 7 | 1033 | 7135573 |