siRNA Details
| Gene Symbol: Capzb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005903.1 | NP_001005903.1 | 3 | 261 | ||
| XM_006239171.2 | XP_006239233.1 | 4 | 1313 | |||
| XM_006239172.2 | XP_006239234.1 | 4 | 1310 | |||
| XM_006239174.2 | XP_006239236.1 | 3 | 1240 | |||
| XM_006239175.2 | XP_006239237.1 | 3 | 1236 | |||
| XM_006239177.2 | XP_006239239.1 | 3 | 290 | |||
| GenBank mRNA | BC083861.1 | - | 3 | 261 | 7126848 | |

Capzb
Genomic Map