siRNA Details
| Gene Symbol: Ccin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005904.1 | NP_001005904.1 | 1 | 664 | ||
| GenBank mRNA | BC083832.1 | - | 1 | 664 | 7112736 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccin| Gene Symbol: Ccin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005904.1 | NP_001005904.1 | 1 | 664 | ||
| GenBank mRNA | BC083832.1 | - | 1 | 664 | 7112736 | |