siRNA Details
| Gene Symbol: VASH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014909.4 | NP_055724.1 | 4 | 1412 | ||
| XM_017021087.1 | XP_016876576.1 | 6 | 487 | |||
| XM_017021088.1 | XP_016876577.1 | 5 | 444 | |||
| XM_017021089.1 | XP_016876578.1 | 5 | 303 | |||
| XM_017021090.1 | XP_016876579.1 | 4 | 236 | |||
| XM_017021091.1 | XP_016876580.1 | 4 | 408 | |||
| GenBank mRNA | AB028959.1 | - | 4 | 867 | ||
| AL832588.2 | - | 4 | 1116 | |||
| BC009031.1 | - | 4 | 975 | 4156063 | ||
| BC051896.1 | - | 4 | 706 | 5170122 | ||

VASH1
Genomic Map