siRNA Details
| Gene Symbol: ZNF507 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136156.1 | NP_001129628.1 | 3 | 585 | ||
| NM_014910.4 | NP_055725.2 | 2 | 491 | |||
| GenBank mRNA | AB029007.1 | - | 3 | 546 | ||
| AK292526.1 | - | 2 | 489 | |||
| AL713775.1 | - | 3 | 472 | |||
| BC136417.1 | - | 3 | 521 | 9020405 | ||
| BX640881.1 | - | 2 | 453 | |||
| DB165165.1 | - | 2 | 491 | |||

ZNF507
Genomic Map