siRNA Details
| Gene Symbol: Best1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011913.2 | NP_036043.2 | 12 | 1972 | ||
| GenBank mRNA | AK006549.1 | - | 10 | 1591 | ||
| AY450427.1 | - | 11 | 1796 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Best1| Gene Symbol: Best1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011913.2 | NP_036043.2 | 12 | 1972 | ||
| GenBank mRNA | AK006549.1 | - | 10 | 1591 | ||
| AY450427.1 | - | 11 | 1796 | |||