siRNA Details
| Gene Symbol: CLEC16A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015226.2 | NP_056041.1 | 24 | 3860 | ||
| XM_005255210.1 | XP_005255267.1 | 23 | 3848 | |||
| XM_005255211.1 | XP_005255268.1 | 24 | 3806 | |||
| XM_005255213.1 | XP_005255270.1 | 23 | 3800 | |||
| XM_005255214.1 | XP_005255271.1 | 23 | 3689 | |||
| XM_011522434.1 | XP_011520736.1 | 23 | 3725 | |||
| XM_011522435.1 | XP_011520737.1 | 25 | 3978 | |||
| XM_017023090.1 | XP_016878579.1 | 12 | 2139 | |||
| GenBank mRNA | AB002348.3 | - | 24 | 3763 | ||
| AK126771.1 | - | 4 | 1192 | |||

CLEC16A
Genomic Map