siRNA Details
| Gene Symbol: WSCD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015253.1 | NP_056068.1 | 3 | 797 | ||
| XM_005256572.1 | XP_005256629.1 | 3 | 885 | |||
| XM_005256573.1 | XP_005256630.1 | 3 | 860 | |||
| XM_011523777.1 | XP_011522079.1 | 3 | 885 | |||
| GenBank mRNA | AK309645.1 | - | 2 | 249 | ||
| AL833730.1 | - | 3 | 797 | |||
| BC009975.2 | - | 3 | 822 | 4110708 | ||

WSCD1
Genomic Map