siRNA Details
| Gene Symbol: Rpa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021582.1 | NP_067593.1 | 5, 6 | 485 | ||
| GenBank mRNA | BC079180.1 | - | 5, 6 | 485 | 7130711 | |
| FQ228766.1 | - | 2, 3 | 114 | |||
| X98490.1 | - | 4, 5 | 391 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rpa2| Gene Symbol: Rpa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021582.1 | NP_067593.1 | 5, 6 | 485 | ||
| GenBank mRNA | BC079180.1 | - | 5, 6 | 485 | 7130711 | |
| FQ228766.1 | - | 2, 3 | 114 | |||
| X98490.1 | - | 4, 5 | 391 | |||