siRNA Details
| Gene Symbol: Rsrc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005525.2 | NP_001005525.1 | 5 | 685 | ||
| GenBank mRNA | AK005213.1 | - | 3 | 416 | ||
| AK012697.1 | - | 6 | 752 | |||
| AK054360.1 | - | 2 | 1255 | |||
| AK084745.1 | - | 6 | 732 | |||
| AK166941.1 | - | 8 | 923 | |||
| BC008229.1 | - | 5 | 643 | |||
| BC132550.1 | - | 3 | 469 | 40130827 | ||
| BC132552.1 | - | 3 | 469 | 40130829 | ||
| BC145430.1 | - | 3 | 468 | 9053988 | ||
| BC145431.1 | - | 3 | 469 | |||
| BC145432.1 | - | 3 | 469 | |||
| CK793427.1 | - | 631 | ||||
| DT902145.1 | - | 1 | 148 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001014128, XM_006249339, XM_006249341, XM_006249338, XM_006249340, and others.

Rsrc2
Genomic Map