siRNA Details
| Gene Symbol: ZNF510 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001314059.1 | NP_001300988.1 | 2, 3 | 736 | ||
| NM_014930.2 | NP_055745.1 | 2, 3 | 347 | |||
| XM_005251808.2 | XP_005251865.1 | 2, 3 | 337 | |||
| XM_011518393.2 | XP_011516695.1 | 2, 3 | 767 | |||
| XM_017014483.1 | XP_016869972.1 | 3, 4 | 399 | |||
| GenBank mRNA | AB023189.1 | - | 1, 2 | 264 | ||
| AK292600.1 | - | 2, 3 | 317 | |||
| BC036676.1 | - | 2, 3 | 734 | 4792684 | ||
| BC058022.1 | - | 2, 3 | 311 | |||
| BC068587.1 | - | 2, 3 | 730 | 4374918 | ||

ZNF510
Genomic Map