siRNA Details
| Gene Symbol: Cgrrf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026832.3 | NP_081108.2 | 6 | 1099 | ||
| GenBank mRNA | AK030755.1 | - | 6 | 1101 | ||
| AK154797.1 | - | 6 | 1175 | |||
| BC037677.1 | - | 6 | 1115 | 3491666 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cgrrf1| Gene Symbol: Cgrrf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026832.3 | NP_081108.2 | 6 | 1099 | ||
| GenBank mRNA | AK030755.1 | - | 6 | 1101 | ||
| AK154797.1 | - | 6 | 1175 | |||
| BC037677.1 | - | 6 | 1115 | 3491666 | ||