siRNA Details
| Gene Symbol: Ggnbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027544.2 | NP_081820.2 | 1 | 556 | ||
| GenBank mRNA | AB250379.1 | - | 1 | 439 | ||
| AK077129.1 | - | 1 | 439 | |||
| AY141192.1 | - | 1 | 556 | |||
| BC049764.1 | - | 1 | 417 | 6774330 | ||

Ggnbp1
Genomic Map