siRNA Details
| Gene Symbol: 0610010F05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027860.2 | NP_082136.2 | 14, 15 | 1888 | ||
| GenBank mRNA | AK029538.1 | - | 14, 15 | 1810 | ||
| AK129454.1 | - | 14, 15 | 1855 | |||
| BC055049.1 | - | 13, 14 | 1761 | |||
| BC065122.1 | - | 15, 16 | 1912 | |||
| BC072622.1 | - | 14, 15 | 1878 | 6821718 | ||
| BC079876.1 | - | 14, 15 | 1874 | 6848269 | ||

0610010F05Rik
Genomic Map