siRNA Details
| Gene Symbol: 0610010F05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027860.2 | NP_082136.2 | 22 | 3014 | ||
| GenBank mRNA | AK002465.1 | - | 8 | 1131 | ||
| AK129454.1 | - | 22 | 2981 | |||
| AK142320.1 | - | 7 | 1040 | |||
| BC065122.1 | - | 23 | 3038 | |||
| BC072622.1 | - | 22 | 3005 | 6821718 | ||
| BC079876.1 | - | 22 | 3000 | 6848269 | ||

0610010F05Rik
Genomic Map