siRNA Details
| Gene Symbol: Utp14a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028276.1 | NP_082552.1 | 12 | 1373 | ||
| GenBank mRNA | AK134784.1 | - | 12 | 1398 | ||
| AK172924.1 | - | 12 | 1373 | |||
| AY316163.1 | - | 12 | 1396 | |||
| BC062165.1 | - | 12 | 1422 | |||
| BC082590.1 | - | 12 | 1399 | 30625504 | ||

Utp14a
Genomic Map