siRNA Details
| Gene Symbol: Utp14a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028276.1 | NP_082552.1 | 12 | 1694 | ||
| GenBank mRNA | AK134784.1 | - | 12 | 1719 | ||
| AK172924.1 | - | 12 | 1694 | |||
| AY316163.1 | - | 12 | 1717 | |||
| BC082590.1 | - | 12 | 1720 | 30625504 | ||

Utp14a
Genomic Map