siRNA Details
| Gene Symbol: Fbrsl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142642.1 | NP_001136114.1 | 19 | 2755 | ||
| NM_028596.2 | NP_082872.2 | 12 | 1799 | |||
| GenBank mRNA | AK031967.1 | - | 12 | 1769 | ||
| AK129388.1 | - | 8 | 3311 | |||
| BC059932.1 | - | 17 | 3541 | |||
| BC069994.1 | - | 7 | 756 | |||
| BC151196.1 | - | 14 | 1764 | 9088084 | ||

Fbrsl1
Genomic Map