siRNA Details
| Gene Symbol: Qtrtd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029128.2 | NP_083404.2 | 8 | 1754 | ||
| GenBank mRNA | BC017628.1 | - | 9 | 2781 | ||
| BC083089.1 | - | 8 | 1734 | 30105859 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Qtrtd1| Gene Symbol: Qtrtd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029128.2 | NP_083404.2 | 8 | 1754 | ||
| GenBank mRNA | BC017628.1 | - | 9 | 2781 | ||
| BC083089.1 | - | 8 | 1734 | 30105859 | ||