siRNA Details
Gene Symbol: Lyrm7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006294.1 | - | 6 | 1138 | ||
BC139403.1 | - | 6 | 939 | 8862425 | ||
BC139406.1 | - | 6 | 939 | 8862428 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Lyrm7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006294.1 | - | 6 | 1138 | ||
BC139403.1 | - | 6 | 939 | 8862425 | ||
BC139406.1 | - | 6 | 939 | 8862428 |